2,3-Dihydrobenzo[b]furan-7-carboxylic acid 97%

Supplier: Ambeed
Warning

A253731-100MG A253731-1G A253731-250MG A253731-5G
77262-064EA 9 USD
77262-064 77262-066 77262-068 77262-070
2,3-Dihydrobenzo[b]furan-7-carboxylic acid 97%
2,3-Dihydrobenzo[b]furan-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈O₃
MDL Number: MFCD00671526
CAS Number: 35700-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR