2-Fluoro-m-anisaldehyde 98%

Supplier: Ambeed
Danger

A253723-1G A253723-25G A253723-5G
77238-356EA 18 USD
77238-356 77238-358 77238-360
2-Fluoro-m-anisaldehyde 98%

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MDL Number: MFCD03411962
CAS Number: 103438-88-6
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR