4,4'-Methylenebis(3-chloro-2,6-diethylaniline) 98%

Supplier: Ambeed
Warning

A253313-25G A253313-5G
77289-096EA 15.45 USD
77289-096 77289-098
4,4'-Methylenebis(3-chloro-2,6-diethylaniline) 98%
4,4'-Methylenebis(3-chloro-2,6-diethylaniline)

Precaución: for Research Use Only

Formula: C₂₁H₂₈Cl₂N₂
MW: 379.3664 g/mol
MDL Number: MFCD00071551
CAS Number: 106246-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR