3-Phenoxyphenylacetonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Phenoxyphenyl)acetonitrile

A253229-100G A253229-1G A253229-25G A253229-5G
77289-088EA 491 USD
77289-088 77289-090 77289-092 77289-094
3-Phenoxyphenylacetonitrile 95%
3-Phenoxyphenylacetonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO
MDL Number: MFCD00016393
CAS Number: 51632-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR