2'-Bromo-5'-fluoroacetophenone 98%

Supplier: Ambeed
Warning

A253156-100G A253156-1G A253156-5G
77262-040EA 339.9 USD
77262-040 77262-042 77262-044
2'-Bromo-5'-fluoroacetophenone 98%
2'-Bromo-5'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD07780635
CAS Number: 1006-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR