Pyridin-2-amine hydrochloride 97%

Supplier: Ambeed
Warning

A252957-1G A252957-250MG A252957-5G
77238-328EA 33 USD
77238-328 77238-330 77238-332
Pyridin-2-amine hydrochloride 97%
Pyridin-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₇ClN₂
MDL Number: MFCD00137029
CAS Number: 32654-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR