4-(1H-Tetrazol-5-yl)benzoic acid 98%

Supplier: Ambeed
Danger

A252956-100MG A252956-1G A252956-250MG A252956-5G
77289-080EA 17.51 USD
77289-080 77289-082 77289-084 77289-086
4-(1H-Tetrazol-5-yl)benzoic acid 98%
4-(1H-Tetrazol-5-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆N₄O₂
MDL Number: MFCD06345720
CAS Number: 34114-12-0
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR