4-Formyl-2-methoxy-3-nitrophenyl acetate 95+%

Supplier: Ambeed
Warning

A252715-100MG A252715-1G A252715-250MG
77289-072EA 28.84 USD
77289-072 77289-074 77289-076
4-Formyl-2-methoxy-3-nitrophenyl acetate 95+%
4-Formyl-2-methoxy-3-nitrophenyl acetate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₆
MW: 239.18 g/mol
MDL Number: MFCD00086004
CAS Number: 2698-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR