Diacetato[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]ruthenium(II) 98%

Supplier: Ambeed
Warning

Synonyms: Ru(OAc)2[(S)-segphos®]

A252714-250MG A252714-5G
77249-978EA 45 USD
77249-978 77249-980
Diacetato[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]ruthenium(II) 98%
Diacetato[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]ruthenium(II)

Precaución: for Research Use Only

Formula: C₄₂H₃₄O₈P₂Ru
MW: 829.7300 g/mol
MDL Number: MFCD09753024
CAS Number: 373650-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR