3-(tert-Butyl)-1-ethyl-1H-pyrazole-5-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A252620-100MG
A252620-250MG
77276-364EA
83.43
USD
77276-364
77276-366
3-(tert-Butyl)-1-ethyl-1H-pyrazole-5-carboxylic acid 97%
3-(tert-Butyl)-1-ethyl-1H-pyrazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₆N₂O₂ MW: 196.24 g/mol |
MDL Number:
MFCD00208046 CAS Number: 195447-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...