3-tert-Butylbenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(tert-Butyl)benzoic acid

A252569-100MG A252569-10G A252569-1G A252569-250MG A252569-5G
77276-354EA 16.48 USD
77276-354 77276-356 77276-358 77276-360 77276-362
3-tert-Butylbenzoic acid 95%
3-tert-Butylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD01463861
CAS Number: 7498-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR