3-Bromo-N-methoxy-N-methylbenzamide 98%

Supplier: Ambeed
Warning

A252486-10G A252486-1G A252486-5G
77276-350EA 132.87 USD
77276-350 77276-352 77748-473
3-Bromo-N-methoxy-N-methylbenzamide 98%
3-Bromo-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.08 g/mol
MDL Number: MFCD02684302
CAS Number: 207681-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR