(2-Bromo-3-pyridinyl)methanol 96%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3-pyridinemethanol, (2-Bromopyridin-3-yl)methanol, 2-Bromo-3-(hydroxymethyl)pyridine

A251893-100MG A251893-1G A251893-250MG A251893-5G
77262-014EA 27.81 USD
77262-014 77262-016 77262-018 77262-020
(2-Bromo-3-pyridinyl)methanol 96%
(2-Bromo-3-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD08276300
CAS Number: 131747-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR