4-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: N-(4-Carboxyphenyl)maleimide, 4-Maleimidobenzoic acid, 4-(2,5-Dioxo-2,5-dihydro-pyrrol-1-yl)-benzoicacid

A251871-1G A251871-250MG A251871-25G
77289-058EA 22.99 USD
77289-058 77289-060 77289-062
4-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoic acid 97%
4-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₇NO₄
MDL Number: MFCD00458571
CAS Number: 17057-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR