2-Bromo-6-iodo-4-(trifluoromethyl)aniline 95%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-bromo-5-iodobenzotrifluoride

A251844-100MG A251844-1G
77262-010EA 37.08 USD
77262-010 77262-012
2-Bromo-6-iodo-4-(trifluoromethyl)aniline 95%
2-Bromo-6-iodo-4-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄BrF₃IN
MW: 365.9170 g/mol
MDL Number: MFCD08458149
CAS Number: 875306-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR