2-Chloro-3-methoxyphenol 98%
Supplier: Ambeed
|
Warning
|
A251721-1G
A251721-250MG
A251721-5G
77261-998EA
25.75
USD
77261-998
77262-000
77262-002
2-Chloro-3-methoxyphenol 98%
2-Chloro-3-methoxyphenol
Precaución: for Research Use Only
Formula:
C₇H₇ClO₂ MW: 158.5823 g/mol |
MDL Number:
MFCD08275690 CAS Number: 72232-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...