TDZD-8 98%

Supplier: Ambeed
Warning

Synonyms: 4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione

A251413-10MG A251413-1MG A251413-50MG A251413-5MG
77289-034EA 29.87 USD
77289-034 77289-036 77289-038 77289-040
TDZD-8 98%
TDZD-8

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂SO₂
MW: 222.3 g/mol
MDL Number: MFCD04973552
CAS Number: 327036-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR