1-Iodododecane 98% stabilised

Supplier: Ambeed
Warning

Synonyms: 1-Dodecyl iodide

Stabilization: Stabilised with copper chip

A251153-100G A251153-5G
77238-292EA 76 USD
77238-292 77238-294
1-Iodododecane 98% stabilised
1-Iodododecane

Precaución: for Research Use Only

Formula: C₁₂H₂₅I
MW: 296.24 g/mol
Boiling Pt: 298
Melting Pt: -3
MDL Number: MFCD00001088
CAS Number: 4292-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR