1-(3,4-Dichlorophenyl)cyclopropanecarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A251124-100MG
A251124-1G
A251124-250MG
77238-286EA
10
USD
77238-286
77238-288
77238-290
1-(3,4-Dichlorophenyl)cyclopropanecarboxylic acid 98%
1-(3,4-Dichlorophenyl)cyclopropanecarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₈Cl₂O₂ |
MDL Number:
MFCD00461541 CAS Number: 342386-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...