D-allo-Threonine 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Allo-Thr-OH, (2R,3R)-2-Amino-3-hydroxybutanoic acid

A250942-100G A250942-1G A250942-250MG A250942-25G A250942-5G A250942-10G
77289-018EA 580.92 USD
77289-018 77289-020 77289-022 77289-024 77289-026 77616-042
D-allo-Threonine 98%
D-allo-Threonine

Precaución: for Research Use Only

Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00004526
CAS Number: 24830-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR