3-Benzyloxy-1-propanol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Propanol, 3-(benzyloxy)-, 3-phenylmethoxy-1-propanol, Trimethylene glycol monobenzyl ether, 3-(Benzyloxy)-1-propanol, 3-(Benzyloxy)propan-1-ol

A250837-100G A250837-10G A250837-1G A250837-25G A250837-500G A250837-5G
77276-330EA 57.68 USD
77276-330 77276-332 77276-334 77276-336 77276-338 77276-340
3-Benzyloxy-1-propanol 98%
3-Benzyloxy-1-propanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
MDL Number: MFCD00029659
CAS Number: 4799-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR