4-(trans-4-Amylcyclohexyl)benzoic acid 98%

Supplier: Ambeed
Warning

A250781-100G A250781-10G A250781-1G A250781-25G A250781-5G
77289-008EA 170 USD
77289-008 77289-010 77289-012 77289-014 77289-016
4-(trans-4-Amylcyclohexyl)benzoic acid 98%
4-(trans-4-Amylcyclohexyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₆O₂
MDL Number: MFCD06208360
CAS Number: 65355-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR