2,4-Thiazolidinedione 97%

Supplier: Ambeed
Warning

A250698-1000G A250698-100G A250698-25G A250698-500G A250698-5G
77288-998EA 65 USD
77288-998 77289-000 77289-002 77289-004 77289-006
2,4-Thiazolidinedione 97%
2,4-Thiazolidinedione

Precaución: for Research Use Only

Formula: C₃H₃NO₂S
MDL Number: MFCD00005478
CAS Number: 2295-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR