Ethyl 1-methyl-1H-pyrazole-5-carboxylate 98%

Supplier: Ambeed
Warning

A250535-1G A250535-5G
77238-282EA 11.33 USD
77238-282 77238-284
Ethyl 1-methyl-1H-pyrazole-5-carboxylate 98%
Ethyl 1-methyl-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.166 g/mol
MDL Number: MFCD08700760
CAS Number: 197079-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR