2-Cyano-3-(thiazol-2-yl)acrylic acid 97%

Supplier: Ambeed
Warning

A250388-1G A250388-250MG
77238-278EA 163.77 USD
77238-278 77238-280
2-Cyano-3-(thiazol-2-yl)acrylic acid 97%
2-Cyano-3-(thiazol-2-yl)acrylic acid

Precaución: for Research Use Only

Formula: C₇H₄N₂O₂S
MW: 180.18 g/mol
MDL Number: MFCD26811022
CAS Number: 1567534-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR