Sodium ((4-aminophenyl)sulfonyl)(6-chloropyrazin-2-yl)amide 99+%

Supplier: Ambeed
Warning

Synonyms: sulphaclozine sodium

A250306-100MG A250306-1MG
77261-982EA 215 USD
77261-982 77748-469
Sodium ((4-aminophenyl)sulfonyl)(6-chloropyrazin-2-yl)amide 99+%
Sodium ((4-aminophenyl)sulfonyl)(6-chloropyrazin-2-yl)amide

Precaución: for Research Use Only

Formula: C₁₀H₈ClN₄NaO₂S
MW: 306.7 g/mol
MDL Number: MFCD06798154
CAS Number: 23307-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR