1H-1,2,4-Triazole-3-thiol 95%

Supplier: Ambeed
Warning

A250059-1000G A250059-100G A250059-25G A250059-500G A250059-5G
77276-316EA 67 USD
77276-316 77276-318 77276-320 77276-322 77276-324
1H-1,2,4-Triazole-3-thiol 95%
1H-1,2,4-Triazole-3-thiol

Precaución: for Research Use Only

Formula: C₂H₃N₃S
MDL Number: MFCD00005229
CAS Number: 3179-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR