3,4-Dichloro-2-fluoropyridine 95%

Supplier: Ambeed
Warning

A250045-100MG A250045-1G A250045-250MG A250045-5G
77276-308EA 39.14 USD
77276-308 77276-310 77276-312 77276-314
3,4-Dichloro-2-fluoropyridine 95%
3,4-Dichloro-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂FN
MW: 165.98 g/mol
MDL Number: MFCD09031167
CAS Number: 851179-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR