3-Bromo-α-methylbenzyl alcohol 98%

Supplier: Ambeed
Danger

Synonyms: 3-Bromophenyl methyl carbinol, m-Bromo-α-methylbenzyl alcohol, 1-(3-Bromophenyl)ethanol

A249971-1G A249971-25G A249971-5G
77276-298EA 9 USD
77276-298 77276-300 77276-302
3-Bromo-α-methylbenzyl alcohol 98%
3-Bromo-α-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06
MDL Number: MFCD00021863
CAS Number: 52780-14-0
UN: 1987
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR