2-Chloro-5-methylphenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (2-Chloro-5-methylphenyl)boronic acid, 2-Chloro-5-methylbenzeneboronic acid

A249805-1G A249805-25G
77276-294EA 11 USD
77276-294 77276-296
2-Chloro-5-methylphenylboronic acid 96%
2-Chloro-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₂
MDL Number: MFCD06659822
CAS Number: 193353-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR