2,5,6-Trifluoronicotinonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Cyano-2,5,6-trifluoropyridine

A249756-1G A249756-250MG A249756-25G
77249-946EA 38.11 USD
77249-946 77249-948 77746-393
2,5,6-Trifluoronicotinonitrile 98%
2,5,6-Trifluoronicotinonitrile

Precaución: for Research Use Only

Formula: C₆HF₃N₂
MW: 158.0800 g/mol
MDL Number: MFCD04973023
CAS Number: 870065-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR