4,4'-Thiobisbenzenethiol 98%
Supplier: Ambeed
|
Danger
|
A249725-1G
A249725-25G
A249725-5G
77288-974EA
10.3
USD
77288-974
77288-976
77288-978
4,4'-Thiobisbenzenethiol 98%
4,4'-Thiobisbenzenethiol
Precaución: for Research Use Only
Formula:
C₁₂H₁₀S₃ MW: 250.4028 g/mol |
MDL Number:
MFCD00142119 CAS Number: 19362-77-7 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...