3,4-Difluorothiophenol 98%

Supplier: Ambeed
Danger

A249639-1G A249639-5G
77276-290EA 14 USD
77276-290 77276-292
3,4-Difluorothiophenol 98%
3,4-Difluorothiophenol

Precaución: for Research Use Only

Formula: C₆H₄F₂S
MDL Number: MFCD03094279
CAS Number: 60811-24-7
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR