3-Amino-5-chlorobenzoic acid 98%

Supplier: Ambeed
Warning

A249530-1G A249530-250MG A249530-5G
77276-284EA 39.14 USD
77276-284 77276-286 77276-288
3-Amino-5-chlorobenzoic acid 98%
3-Amino-5-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.5810 g/mol
MDL Number: MFCD02183894
CAS Number: 21961-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR