2-Biphenylylmethanol 95%

Supplier: Ambeed
Warning

Synonyms: 2-Biphenylmethanol, 2-Phenylbenzyl alcohol, [1,1-Biphenyl]-2-ylmethanol

A249381-1G A249381-5G
77261-968EA 29 USD
77261-968 77261-970
2-Biphenylylmethanol 95%
2-Biphenylylmethanol

Precaución: for Research Use Only

Formula: C₁₃H₁₂O
MW: 184.24 g/mol
Boiling Pt: 335
MDL Number: MFCD00004620
CAS Number: 2928-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR