2-Methyl-2H-indazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Carboxy-2-methyl-2H-indazole

A249211-10G A249211-250MG A249211-5G A249211-25G A249211-1G
77261-954EA 91.67 USD
77261-954 77261-956 77261-958 77616-038 77748-463
2-Methyl-2H-indazole-3-carboxylic acid 95%
2-Methyl-2H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.172 g/mol
MDL Number: MFCD07368556
CAS Number: 34252-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR