3-(Trifluoromethylthio)benzaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 3-((Trifluoromethyl)thio)benzaldehyde

A249032-10G A249032-1G A249032-250MG A249032-5G A249032-100MG
77276-278EA 201 USD
77276-278 77276-280 77276-282 77593-570 77748-462
3-(Trifluoromethylthio)benzaldehyde 95%
3-(Trifluoromethylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅F₃OS
MDL Number: MFCD00236335
CAS Number: 51748-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR