6-Nitro-o-toluic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-6-nitrobenzoic acid

A248932-100G A248932-1G A248932-25G A248932-10G
77238-226EA 173.04 USD
77238-226 77238-228 77238-230 77746-392
6-Nitro-o-toluic acid 98%
6-Nitro-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MW: 181.15 g/mol
MDL Number: MFCD00007267
CAS Number: 13506-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR