AST-487 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-((4-Ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)-3-(4-((6-(methylamino)pyrimidin-4-yl)oxy)phenyl)urea

A248926-10MG A248926-1MG A248926-5MG
77238-220EA 364.62 USD
77238-220 77238-222 77238-224
AST-487 98%
AST-487

Precaución: for Research Use Only

Formula: C₂₆H₃₀F₃N₇O₂
MW: 529.6 g/mol
MDL Number: MFCD11983171
CAS Number: 630124-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR