4-Methyl-2,2'-bipyridine 98%

Supplier: Ambeed
Warning

A248823-100MG A248823-1G A248823-250MG
77288-960EA 24 USD
77288-960 77288-962 77288-964
4-Methyl-2,2'-bipyridine 98%
4-Methyl-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂
MW: 170.21 g/mol
MDL Number: MFCD08062570
CAS Number: 56100-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR