3,5-Dimethyl-4-nitroisoxazole 97%

Supplier: Ambeed
Warning

A248390-1G A248390-25G A248390-5G
77276-250EA 27.81 USD
77276-250 77276-252 77276-254 77276-256
3,5-Dimethyl-4-nitroisoxazole 97%
3,5-Dimethyl-4-nitroisoxazole

Precaución: for Research Use Only

Formula: C₅H₆N₂O₃
MDL Number: MFCD00051656
CAS Number: 1123-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR