2-(5-(tert-Butyl)isoxazol-3-yl)-N-(3-chlorophenyl)-2-oxoacetohydrazonoyl cyanide 98%

Supplier: Ambeed
Warning

A248229-10MG A248229-1MG A248229-5MG
77238-204EA 91.67 USD
77238-204 77238-206 77238-208
2-(5-(tert-Butyl)isoxazol-3-yl)-N-(3-chlorophenyl)-2-oxoacetohydrazonoyl cyanide 98%
2-(5-(tert-Butyl)isoxazol-3-yl)-N-(3-chlorophenyl)-2-oxoacetohydrazonoyl cyanide

Precaución: for Research Use Only

Formula: C₁₆H₁₅ClN₄O₂
MW: 330.77 g/mol
CAS Number: 263707-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR