4-Chlorophenyl sulfoxide 95%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Dichlordiphenylsulphoxid, Bis(4-chlorophenyl) Sulphoxide

A248145-100G A248145-25G A248145-5G
77288-940EA 59 USD
77288-940 77288-942 77288-944
4-Chlorophenyl sulfoxide 95%
4-Chlorophenyl sulfoxide

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂OS
MDL Number: MFCD00000618
CAS Number: 3085-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR