2,2'-(9H-fluorene-9,9-diyl)diethanol 97%

Supplier: Ambeed
Warning

A247975-100G A247975-10G A247975-1G A247975-250MG A247975-25G A247975-5G
77249-902EA 865 USD
77249-902 77249-904 77249-906 77249-908 77249-910 77249-912
2,2'-(9H-fluorene-9,9-diyl)diethanol 97%
2,2'-(9H-fluorene-9,9-diyl)diethanol

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₂
MDL Number: MFCD20488038
CAS Number: 203070-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR