3,3'-Diaminobenzidine (DAB) 98%

Supplier: Ambeed
Danger

A247767-1G A247767-25G A247767-5G A247767-100G A247767-500G
77276-236EA 22 USD
77276-236 77276-238 77276-240 77748-452 77748-453
3,3'-Diaminobenzidine (DAB) 98%
3,3'-Diaminobenzidine (DAB)

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₄
MDL Number: MFCD00007725
CAS Number: 91-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR