Methyl-3-formyl-1H-indole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-formyl-1H-indole-4-carboxylate

A247532-100MG A247532-10G A247532-1G A247532-250MG A247532-5G
77276-226EA 16.48 USD
77276-226 77276-228 77276-230 77276-232 77276-234
Methyl-3-formyl-1H-indole-4-carboxylate 97%
Methyl-3-formyl-1H-indole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MW: 203.2 g/mol
MDL Number: MFCD06656838
CAS Number: 53462-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR