2-Hydrazino-1H-1,3-benzimidazole 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Hydrazinyl-1H-benzo[d]imidazole
A247519-10G
A247519-1G
A247519-250MG
A247519-5G
77261-938EA
94.76
USD
77261-938
77261-940
77261-942
77261-944
2-Hydrazino-1H-1,3-benzimidazole 95%
2-Hydrazino-1H-1,3-benzimidazole
Precaución: for Research Use Only
Formula:
C₇H₈N₄ MW: 148.17 g/mol |
MDL Number:
MFCD00513603 CAS Number: 15108-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...