3,3',4,4'-Diphenylsulfonetetracarboxylic dianhydride 98%

Supplier: Ambeed
Warning

Synonyms: 5,5'-sulphonylbis(isobenzofuran-1,3-dione), 4,4'-Sulphonyldiphthalic Anhydride

A247411-1G A247411-25G A247411-5G
77276-220EA 13.39 USD
77276-220 77276-222 77276-224
3,3',4,4'-Diphenylsulfonetetracarboxylic dianhydride 98%
3,3',4,4'-Diphenylsulfonetetracarboxylic dianhydride

Precaución: for Research Use Only

Formula: C₁₆H₆O₈S
MW: 358.28 g/mol
MDL Number: MFCD00039145
CAS Number: 2540-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR