1-(3,6-Dibromo-9H-carbazol-9-yl)-3-(piperazin-1-yl)propan-2-ol 97%

Supplier: Ambeed
Warning

Synonyms: BAI1

A247314-1MG A247314-5MG A247314-100MG A247314-10MG A247314-250MG A247314-50MG
77276-216EA 15.45 USD
77276-216 77276-218 77748-448 77748-449 77748-450 77748-451
1-(3,6-Dibromo-9H-carbazol-9-yl)-3-(piperazin-1-yl)propan-2-ol 97%
1-(3,6-Dibromo-9H-carbazol-9-yl)-3-(piperazin-1-yl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₉H₂₁Br₂N₃O
MW: 467.19 g/mol
MDL Number: MFCD00687415
CAS Number: 335165-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR