6-Bromo-2,3-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A247308-10G A247308-1G A247308-250MG
77249-896EA 287 USD
77249-896 77249-898 77249-900
6-Bromo-2,3-difluorobenzoic acid 97%
6-Bromo-2,3-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O₂
MDL Number: MFCD07368092
CAS Number: 183065-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR